Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adh6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106475.1 | NP_001099945.1 | 3 - 4 | 459 | 116 | |
XM_006233371.2 | XP_006233433.1 | 4 - 5 | 541 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |