Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrriq3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024307.1 | NP_001019478.1 | 1 - 2 | 139 | 71 | |
GenBank mRNA | BC078994.1 | - | 1 - 2 | 139 | 7112662 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |