Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF063102.1 | - | 20 - 21 | 3374 | 100 | |
AF081148.1 | - | 19 - 20 | 3748 | 100 | ||
AF081149.1 | - | 19 - 20 | 3748 | 100 | ||
AF081150.1 | - | 19 - 20 | 3748 | 100 | ||
AF081151.1 | - | 20 - 21 | 3793 | 100 | ||
AF081152.1 | - | 20 - 21 | 3793 | 100 | ||
AF081153.1 | - | 20 - 21 | 3793 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |