Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lphn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF063102.1 | - | 9 - 10 | 1855 | 98 | |
AF081148.1 | - | 10 - 11 | 2301 | 98 | ||
AF081149.1 | - | 10 - 11 | 2301 | 98 | ||
AF081150.1 | - | 10 - 11 | 2301 | 98 | ||
AF081151.1 | - | 10 - 11 | 2301 | 98 | ||
AF081152.1 | - | 10 - 11 | 2301 | 98 | ||
AF081153.1 | - | 10 - 11 | 2301 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |