Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107714.1 | NP_001101184.1 | 8 - 9 | 1046 | 106 | |
XM_006233114.2 | XP_006233176.1 | 8 - 9 | 995 | 106 | ||
XM_008761396.1 | XP_008759618.1 | 7 - 8 | 1442 | 106 | ||
XM_008761397.1 | XP_008759619.1 | 7 - 8 | 1442 | 106 | ||
XM_008761398.1 | XP_008759620.1 | 8 - 9 | 995 | 106 | ||
XM_008761399.1 | XP_008759621.1 | 8 - 9 | 1079 | 106 | ||
GenBank mRNA | BC158880.1 | - | 8 - 9 | 982 | 7936253 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |