Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sypl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108563.1 | NP_001102033.1 | 4 - 5 | 638 | 77 | |
GenBank mRNA | AB158471.1 | - | 4 - 5 | 456 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |