Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772103.1 | XP_008770325.1 | 14 - 15 | 1887 | 78 | |
XM_008772104.1 | XP_008770326.1 | 14 - 15 | 1887 | 78 | ||
GenBank mRNA | BC098944.1 | - | 7 - 8 | 835 | 78 | |
BC168151.1 | - | 14 - 15 | 1816 | 7459379 | 78 | |
CO558582.1 | - | 1 - 2 | 38 | 78 | ||
DV722630.1 | - | 4 - 5 | 386 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |