Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106810.1 | NP_001100280.1 | 4 - 5 | 1868 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Recommended Assay Rattus norvegicus Probe spans exons |