Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rai14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011947.1 | NP_001011947.1 | 13 - 14 | 1130 | 86 | |
XM_006232060.2 | XP_006232122.1 | 14 - 15 | 1704 | 86 | ||
XM_006232061.2 | XP_006232123.1 | 14 - 15 | 1277 | 86 | ||
XM_006232062.2 | XP_006232124.1 | 14 - 15 | 2142 | 86 | ||
XM_006232063.2 | XP_006232125.1 | 13 - 14 | 1617 | 86 | ||
XM_006232064.2 | XP_006232126.1 | 12 - 13 | 1530 | 86 | ||
XM_008760765.1 | XP_008758987.1 | 12 - 13 | 2073 | 86 | ||
GenBank mRNA | BC085775.1 | - | 13 - 14 | 1130 | 7096978 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |