Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105941.2 | NP_001099411.2 | 1 - 2 | 146 | 72 | |
GenBank mRNA | CB613801.1 | - | 1 - 2 | 152 | 72 | |
FQ217708.1 | - | 1 - 2 | 159 | 72 | ||
FQ221841.1 | - | 1 - 2 | 106 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Inventoried Probe spans exons |