Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsbg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080096.1 | NP_001073565.1 | 13 - 14 | 2197 | 131 | |
XM_006244311.2 | XP_006244373.1 | 13 - 14 | 2706 | 131 | ||
GenBank mRNA | BC129110.1 | - | 13 - 14 | 2197 | 7111052 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |