Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khsrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133602.1 | NP_598286.1 | 15 - 16 | 1690 | 139 | |
XM_006244292.2 | XP_006244354.1 | 15 - 16 | 1828 | 139 | ||
GenBank mRNA | AF308818.1 | - | 15 - 16 | 1690 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |