Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099519.1 | NP_001092989.1 | 5 - 6 | 1648 | 72 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Probe spans exons |