Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109344.2 | NP_001102814.1 | 5 - 6 | 672 | 83 | |
XM_006235673.2 | XP_006235735.1 | 5 - 6 | 851 | 83 | ||
GenBank mRNA | BC167027.1 | - | 5 - 6 | 672 | 7315978 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus Probe spans exons |