Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033708.1 | NP_001028880.1 | 10 - 11 | 928 | 65 | |
XM_006235692.2 | XP_006235754.1 | 11 - 12 | 1239 | 65 | ||
XM_008762517.1 | XP_008760739.1 | 9 - 10 | 1530 | 65 | ||
XM_008762518.1 | XP_008760740.1 | 10 - 11 | 2122 | 65 | ||
XM_008762519.1 | XP_008760741.1 | 10 - 11 | 852 | 65 | ||
GenBank mRNA | BC104721.1 | - | 10 - 11 | 928 | 7383476 | 65 |
BC105758.1 | - | 10 - 11 | 935 | 5623260 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |