Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198780.3 | NP_942075.1 | 7 - 8 | 1331 | 91 | |
GenBank mRNA | BC081900.1 | - | 7 - 8 | 1304 | 7110760 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |