Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pa2g4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004206.1 | NP_001004206.1 | 5 - 6 | 607 | 63 | |
GenBank mRNA | BC079095.1 | - | 5 - 6 | 607 | 7124036 | 63 |
FQ233061.1 | - | 5 - 6 | 615 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |