Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173130.1 | NP_775153.1 | 4 - 5 | 1586 | 103 | |
XM_006228015.1 | XP_006228077.1 | 4 - 5 | 1588 | 103 | ||
GenBank mRNA | AF016182.1 | - | 4 - 5 | 1586 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Rattus norvegicus Probe spans exons |