Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024140.2 | NP_077054.1 | 3 - 4 | 504 | 66 | |
GenBank mRNA | FQ213074.1 | - | 3 - 4 | 302 | 66 | |
L09119.1 | - | 3 - 4 | 504 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |