Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022701.2 | NP_073192.2 | 11 - 12 | 1262 | 95 | |
XM_006256024.2 | XP_006256086.1 | 11 - 12 | 1281 | 95 | ||
GenBank mRNA | BC064652.1 | - | 11 - 12 | 1261 | 5598180 | 95 |
CB324456.1 | - | 2 - 3 | 95 | |||
CB326388.1 | - | 2 - 3 | 95 | |||
U60976.1 | - | 11 - 12 | 1192 | 95 | ||
U60977.1 | - | 9 - 10 | 1689 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |