Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109883.2 | NP_001103353.1 | 14 - 15 | 1406 | 56 | |
GenBank mRNA | AF293383.1 | - | 13 - 14 | 1302 | 56 | |
CB801303.1 | - | 2 - 3 | 219 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Detect Genomic DNA Rattus norvegicus Probe spans exons |