Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166022.1 | NP_001159494.1 | 5 - 6 | 672 | 134 | |
NM_013137.2 | NP_037269.2 | 5 - 6 | 759 | 134 | ||
GenBank mRNA | BC166604.1 | - | 5 - 6 | 759 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |