Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750414.3 | XP_003750462.1 | 1 - 2 | 818 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |