Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Endou | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191069.1 | NP_001177998.1 | 6 - 7 | 751 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |