Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106795.1 | NP_001100265.1 | 11 - 12 | 1233 | 127 | |
XM_006242387.2 | XP_006242449.1 | 10 - 11 | 1150 | 127 | ||
XM_008765704.1 | XP_008763926.1 | 10 - 11 | 1193 | 127 | ||
XM_008765705.1 | XP_008763927.1 | 10 - 11 | 1174 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |