Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Letmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122781.3 | NP_001116253.3 | 6 - 7 | 795 | 62 | |
XM_006242354.2 | XP_006242416.1 | 6 - 7 | 858 | 62 | ||
GenBank mRNA | BC159426.1 | - | 6 - 7 | 776 | 9034024 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons |