Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setdb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271175.1 | NP_001258104.1 | 19 - 20 | 3596 | 58 | |
XM_006232913.2 | XP_006232975.1 | 20 - 21 | 3986 | 58 | ||
XM_006232914.2 | XP_006232976.1 | 19 - 20 | 3767 | 58 | ||
XM_006232916.2 | XP_006232978.1 | 15 - 16 | 2963 | 58 | ||
XM_006232917.2 | XP_006232979.1 | 15 - 16 | 3466 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |