Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191569.1 | NP_001178498.1 | 5 - 6 | 1496 | 118 | |
XM_006233069.1 | XP_006233131.1 | 5 - 6 | 1487 | 118 | ||
XM_006233070.2 | XP_006233132.1 | 5 - 6 | 1814 | 118 | ||
XM_006233071.2 | XP_006233133.1 | 5 - 6 | 1599 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |