Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012081.1 | NP_001012081.1 | 7 - 8 | 1235 | 140 | |
XM_006232967.1 | XP_006233029.1 | 7 - 8 | 1442 | 140 | ||
XM_008761310.1 | XP_008759532.1 | 7 - 8 | 1165 | 140 | ||
XM_008761311.1 | XP_008759533.1 | 7 - 8 | 1442 | 140 | ||
XM_008761312.1 | XP_008759534.1 | 6 - 7 | 1256 | 140 | ||
GenBank mRNA | BC083667.1 | - | 7 - 8 | 1235 | 7135563 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |