Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022589.1 | NP_072111.1 | 4 - 5 | 370 | 80 | |
GenBank mRNA | AJ271442.1 | - | 4 - 5 | 370 | 80 | |
BC085733.1 | - | 4 - 5 | 390 | 7122828 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |