Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109655.2 | NP_001103125.1 | 10 - 11 | 950 | 93 | |
XM_006232988.2 | XP_006233050.1 | 11 - 12 | 1495 | 93 | ||
XM_006232989.2 | XP_006233051.1 | 11 - 12 | 1473 | 93 | ||
XM_006232990.2 | XP_006233052.1 | 11 - 12 | 1455 | 93 | ||
GenBank mRNA | EF187253.1 | - | 10 - 11 | 950 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |