Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pinx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083337.1 | NP_001076806.1 | 3 - 4 | 227 | 131 | |
XM_006252181.2 | XP_006252243.1 | 3 - 4 | 1130 | 131 | ||
GenBank mRNA | BC158553.1 | - | 3 - 4 | 326 | 9088212 | 131 |
EF446165.1 | - | 3 - 4 | 227 | 131 | ||
FQ232228.1 | - | 2 - 3 | 115 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |