Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006993.1 | NP_001006994.1 | 5 - 6 | 593 | 66 | |
GenBank mRNA | BC083748.1 | - | 5 - 6 | 593 | 7192349 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |