Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa2f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080775.1 | NP_542953.1 | 3 - 4 | 327 | 118 | |
XM_008762380.1 | XP_008760602.1 | 3 - 4 | 439 | 118 | ||
GenBank mRNA | M83210.1 | - | 3 - 4 | 327 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |