Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235401.1 | XP_006235463.1 | 1 - 2 | 107 | 60 | |
GenBank mRNA | BC158548.1 | - | 1 - 2 | 70 | 9036351 | 60 |
FQ215756.1 | - | 1 - 2 | 44 | 60 | ||
FQ219977.1 | - | 1 - 2 | 48 | 60 | ||
FQ220245.1 | - | 1 - 2 | 48 | 60 | ||
FQ220303.1 | - | 1 - 2 | 48 | 60 | ||
FQ220687.1 | - | 1 - 2 | 50 | 60 | ||
FQ227515.1 | - | 1 - 2 | 51 | 60 | ||
FQ229420.1 | - | 1 - 2 | 48 | 60 | ||
FQ229727.1 | - | 1 - 2 | 51 | 60 | ||
FQ229762.1 | - | 1 - 2 | 50 | 60 | ||
FQ229855.1 | - | 1 - 2 | 84 | 60 | ||
FQ230056.1 | - | 1 - 2 | 51 | 60 | ||
FQ230407.1 | - | 1 - 2 | 64 | 60 | ||
FQ231493.1 | - | 1 - 2 | 64 | 60 | ||
FQ232091.1 | - | 1 - 2 | 44 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |