Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004230.1 | NP_001004230.1 | 7 - 8 | 902 | 110 | |
GenBank mRNA | BC079029.1 | - | 7 - 8 | 902 | 7114041 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |