Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053393.2 | NP_445845.2 | 3 - 4 | 1079 | 114 | |
XM_008772244.1 | XP_008770466.1 | 2 - 3 | 426 | 114 | ||
GenBank mRNA | AB010436.1 | - | 3 - 4 | 1079 | 114 | |
AB010437.1 | - | 3 - 4 | 1079 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |