Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256466.1 | NP_001243395.1 | 4 - 5 | 508 | 58 | |
XM_008772319.1 | XP_008770541.1 | 5 - 6 | 685 | 58 | ||
GenBank mRNA | AM747280.1 | - | 4 - 5 | 508 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |