Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035257.1 | NP_001030334.1 | 3 - 4 | 365 | 55 | |
GenBank mRNA | BC079370.1 | - | 3 - 4 | 351 | 55 | |
BC104702.1 | - | 3 - 4 | 351 | 55 | ||
BC127449.1 | - | 3 - 4 | 366 | 8365327 | 55 | |
CK600220.1 | - | 3 - 4 | 385 | 55 | ||
FQ212517.1 | - | 3 - 4 | 370 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |