Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109520.1 | NP_001102990.1 | 2 - 3 | 343 | 87 | |
XM_006251660.2 | XP_006251722.1 | 3 - 4 | 476 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |