Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105718.2 | NP_001099188.2 | 12 - 13 | 1579 | 79 | |
GenBank mRNA | BC169004.1 | - | 7 - 8 | 1020 | 79 | |
CK596387.1 | - | 1 - 2 | 144 | 79 | ||
CV120410.1 | - | 4 - 5 | 558 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |