Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pccb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017030.2 | NP_058726.2 | 13 - 14 | 1409 | 79 | |
GenBank mRNA | BC078855.1 | - | 13 - 14 | 1409 | 7108495 | 79 |
M14634.1 | - | 13 - 14 | 1410 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |