Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031642.4 | NP_113830.2 | 3 - 4 | 1044 | 88 | |
GenBank mRNA | AF001417.1 | - | 3 - 4 | 824 | 88 | |
BC097306.1 | - | 3 - 4 | 969 | 7444817 | 88 | |
FQ225029.1 | - | 2 - 3 | 600 | 88 | ||
FQ231202.1 | - | 2 - 3 | 384 | 88 | ||
FQ231451.1 | - | 2 - 3 | 384 | 88 | ||
FQ232231.1 | - | 2 - 3 | 383 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |