Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053462.2 | NP_445914.2 | 679 | 126 | ||
XM_006235365.2 | XP_006235427.1 | 678 | 126 | |||
GenBank mRNA | AF336041.1 | - | 640 | 126 | ||
BC089205.1 | - | 645 | 126 | |||
BC104699.1 | - | 679 | 7189705 | 126 |
Gene Symbol: LOC100911034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749609.2 | XP_003749657.1 | 6 - 7 | 684 | 126 | |
XM_006235451.2 | XP_006235513.1 | 6 - 7 | 678 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |