Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blvra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053850.1 | NP_446302.1 | 1 - 2 | 108 | 92 | |
XM_006234894.2 | XP_006234956.1 | 2 - 3 | 248 | 94 | ||
XM_006234895.2 | XP_006234957.1 | 2 - 3 | 233 | 94 | ||
GenBank mRNA | BC078766.1 | - | 1 - 2 | 120 | 7126157 | 94 |
FQ213399.1 | - | 2 - 3 | 187 | 94 | ||
FQ230935.1 | - | 1 - 2 | 67 | 94 | ||
M81681.1 | - | 1 - 2 | 108 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |