Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153624.2 | NP_705888.2 | 25 | 3944 | 69 | |
XM_006254403.2 | XP_006254465.1 | 25 | 4000 | 69 | ||
GenBank mRNA | AB086193.1 | - | 1 | 53 | 69 | |
BC081716.1 | - | 2 | 236 | 69 | ||
BC166779.1 | - | 7 | 982 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Both primers and probe map within a single exon |