Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myd88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198130.1 | NP_937763.1 | 3 - 4 | 732 | 64 | |
XM_006244087.2 | XP_006244149.1 | 4 - 5 | 1079 | 64 | ||
GenBank mRNA | AY191270.1 | - | 3 - 4 | 732 | 64 | |
BC097266.1 | - | 3 - 4 | 672 | 7459961 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |