Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199490.1 | NP_955784.1 | 2 - 3 | 84 | 69 | |
GenBank mRNA | AY314982.1 | - | 2 - 3 | 84 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |