Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201271.1 | NP_958428.1 | 3 - 4 | 441 | 96 | |
GenBank mRNA | AY299399.1 | - | 3 - 4 | 441 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |