Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201419.1 | NP_958822.1 | 1 - 2 | 129 | 82 | |
GenBank mRNA | AY333961.1 | - | 1 - 2 | 129 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |